COM448618769 Combi-Blocks, Inc.

4-(1-Carboxycyclopropyl)phenylboronic acid, pinacol ester

  • MDL number MFCD09037838
  • Cas Number : 1218790-98-7
  • Molecular Weight : 288.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2346
Availability details for
Quantity
  • Availability
  • Add to Cart