COM448618801 Combi-Blocks, Inc.

3-Hydroxymethyl-pyridyl-5-boronic acid pinacol ester

  • MDL number MFCD11617919
  • Cas Number : 877149-81-0
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2483
Availability details for
Quantity
  • Availability
  • Add to Cart