COM448618824 Combi-Blocks, Inc.

2-(1H-Imidazol-1-yl)pyrimidine-5-boronic acid, pinacol ester

  • MDL number MFCD11878350
  • Cas Number : 1160790-26-0
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2530
Availability details for
Quantity
  • Availability
  • Add to Cart