COM448618830 Combi-Blocks, Inc.

1,3-Dimethyluracil-5-boronic acid pinacol ester

  • MDL number MFCD07781230
  • Cas Number : 269410-01-7
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2654
Availability details for
Quantity
  • Availability
  • Add to Cart