COM448618838 Combi-Blocks, Inc.

2,3-Dimethyl-4-methoxyphenylboronic acid, pinacol ester

  • MDL number MFCD08706293
  • Cas Number : 915402-04-9
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2755
Availability details for
Quantity
  • Availability
  • Add to Cart