COM448618847 Combi-Blocks, Inc.

3-(2-Chloroethoxy)phenylboronic acid, pinacol ester

  • MDL number MFCD09746198
  • Cas Number : 1256359-00-8
  • Molecular Weight : 282.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2801
Availability details for
Quantity
  • Availability
  • Add to Cart