COM448618849 Combi-Blocks, Inc.

2-(2-Chloroethoxy)phenylboronic acid, pinacol ester

  • MDL number MFCD09746200
  • Cas Number : 1256359-02-0
  • Molecular Weight : 282.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2803
Availability details for
Quantity
  • Availability
  • Add to Cart