COM448618860 Combi-Blocks, Inc.

5'-Bromo-2,2'-bithiophene-5-boronic acid pinacol ester

  • MDL number MFCD10566882
  • Cas Number : 578715-23-8
  • Molecular Weight : 371.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2851

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2851
Availability details for
Quantity
  • Availability
  • Add to Cart