COM448618866 Combi-Blocks, Inc.

2-Methylthiazole-5-boronic acid, pinacol ester

  • MDL number MFCD10697442
  • Cas Number : 1218791-01-5
  • Molecular Weight : 225.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2871
Availability details for
Quantity
  • Availability
  • Add to Cart