COM448618880 Combi-Blocks, Inc.

2,6-Dichloro-3-methylpyridine-4-boronic acid pinacol ester

  • MDL number MFCD11617878
  • Cas Number : 1010101-08-2
  • Molecular Weight : 288.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3120

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3120
Availability details for
Quantity
  • Availability
  • Add to Cart