COM448618886 Combi-Blocks, Inc.

5-(Methoxycarbonyl)furan-2-boronic acid pinacol ester

  • MDL number MFCD11846466
  • Cas Number : 676501-87-4
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3158
Availability details for
Quantity
  • Availability
  • Add to Cart