COM448618893 Combi-Blocks, Inc.

1-Methylindole-3-boronic acid pinacol ester

  • MDL number MFCD11894338
  • Cas Number : 683229-61-0
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3360
Availability details for
Quantity
  • Availability
  • Add to Cart