COM448618900 Combi-Blocks, Inc.

2-(1H-Pyrazol-1-yl)pyridine-5-boronic acid, pinacol ester

  • MDL number MFCD11878223
  • Cas Number : 1218791-02-6
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3495
Availability details for
Quantity
  • Availability
  • Add to Cart