COM448618909 Combi-Blocks, Inc.

1-(Toluene-4-sulfonyl)-1h-pyrrole-3-boronic acid pinacol ester

  • MDL number MFCD08063116
  • Cas Number : 1218791-03-7
  • Molecular Weight : 347.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3596
Availability details for
Quantity
  • Availability
  • Add to Cart