COM448618915 Combi-Blocks, Inc.

4-(Methoxycarbonyl)-3-nitrophenylboronic acid, pinacol ester

  • MDL number MFCD16996357
  • Cas Number : 1402238-34-9
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3845
Availability details for
Quantity
  • Availability
  • Add to Cart