COM448618919 Combi-Blocks, Inc.

2-[4-(2-Ethoxyethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD21609556
  • Cas Number : 2096997-06-5
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3920
Availability details for
Quantity
  • Availability
  • Add to Cart