COM448618921 Combi-Blocks, Inc.

4-Chloro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, HCl

  • MDL number MFCD27935904
  • Cas Number : 2377610-52-9
  • Molecular Weight : 290.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-3971
Availability details for
Quantity
  • Availability
  • Add to Cart