COM448618932 Combi-Blocks, Inc.

2-Chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylic acid

  • MDL number MFCD22421502
  • Molecular Weight : 283.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4322
Availability details for
Quantity
  • Availability
  • Add to Cart