COM448618933 Combi-Blocks, Inc.

6-methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD22574004
  • Cas Number : 1375708-04-5
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4370

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4370
Availability details for
Quantity
  • Availability
  • Add to Cart