COM448618942 Combi-Blocks, Inc.

4-(N-Diethylamino)methyl-3-fluorophenylboronic acid pinacol ester

  • MDL number MFCD27981304
  • Cas Number : 2377606-38-5
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4520
Availability details for
Quantity
  • Availability
  • Add to Cart