COM448618953 Combi-Blocks, Inc.

2,3-Difluoro-4-trifluoromethylphenylboronic acid, pinacol ester

  • MDL number MFCD18434440
  • Cas Number : 881402-15-9
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-4783
Availability details for
Quantity
  • Availability
  • Add to Cart