COM448618961 Combi-Blocks, Inc.

3-(3-Chlorophenoxymethyl)phenylboronic acid pinacol ester

  • MDL number MFCD20491435
  • Cas Number : 2096332-64-6
  • Molecular Weight : 344.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-5385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-5385
Availability details for
Quantity
  • Availability
  • Add to Cart