COM448618968 Combi-Blocks, Inc.

2-Hydroxymethylpyridine-4-boronic acid pinacol ester

  • MDL number MFCD12025437
  • Cas Number : 1314135-84-6
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]methanol
Inchi Code 1S/C12H18BNO3/c1-11(2)12(3,4)17-13(16-11)9-5-6-14-10(7-9)8-15/h5-7,15H,8H2,1-4H3
Inchi Key QEOGMZZLSXEKIW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-5835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-5835
Availability details for
Quantity
  • Availability
  • Add to Cart