COM448618978 Combi-Blocks, Inc.

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD12405369
  • Cas Number : 479411-95-5
  • Molecular Weight : 297.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile
Inchi Code 1S/C14H15BF3NO2/c1-12(2)13(3,4)21-15(20-12)11-6-9(8-19)5-10(7-11)14(16,17)18/h5-7H,1-4H3
Inchi Key PDRFNTUDMWZDSL-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6212
Availability details for
Quantity
  • Availability
  • Add to Cart