COM448618979 Combi-Blocks, Inc.

2-(4-Bromofuran-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD12405449
  • Cas Number : 1025719-11-2
  • Molecular Weight : 272.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6228
Availability details for
Quantity
  • Availability
  • Add to Cart