COM448618984 Combi-Blocks, Inc.

5-Chloro-4-methylthiophen-2-boronic acid, pinacol ester

  • MDL number MFCD12405478
  • Cas Number : 1111095-98-7
  • Molecular Weight : 258.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6263
Availability details for
Quantity
  • Availability
  • Add to Cart