COM448618990 Combi-Blocks, Inc.

2,4,6-Trifluorobenzene-1,3,5-triboronic acid, pinacol ester

  • MDL number MFCD12407207
  • Cas Number : 408492-24-0
  • Molecular Weight : 510.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6321
Availability details for
Quantity
  • Availability
  • Add to Cart