COM448618994 Combi-Blocks, Inc.

1-BOC-5-bromopyrrolo[2,3-b]pyridine-3-boronic acid, pinacol ester

  • MDL number MFCD12407269
  • Cas Number : 1025719-14-5
  • Molecular Weight : 423.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6358
Availability details for
Quantity
  • Availability
  • Add to Cart