COM448618996 Combi-Blocks, Inc.

1-Boc-3-methylpyrazole-4-boronic acid pinacol ester

  • MDL number MFCD12407274
  • Cas Number : 1009071-34-4
  • Molecular Weight : 308.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
Inchi Code 1S/C15H25BN2O4/c1-10-11(16-21-14(5,6)15(7,8)22-16)9-18(17-10)12(19)20-13(2,3)4/h9H,1-8H3
Inchi Key CQZGWEHXOJJYPA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6371
Availability details for
Quantity
  • Availability
  • Add to Cart