COM448618998 Combi-Blocks, Inc.

4-[(3-(1H-Imidazol-1-yl)propyl)aminomethyl]phenylboronic acid pinacol ester

  • MDL number MFCD12546046
  • Cas Number : 1257648-78-4
  • Molecular Weight : 341.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6388
Availability details for
Quantity
  • Availability
  • Add to Cart