COM448619023 Combi-Blocks, Inc.

4,5-Difluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester

  • MDL number MFCD13176531
  • Cas Number : 1333122-75-0
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6665

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6665
Availability details for
Quantity
  • Availability
  • Add to Cart