COM448619029 Combi-Blocks, Inc.

1-Methyl-2-oxopyridine-3-boronic acid, pinacol ester

  • MDL number MFCD13182002
  • Cas Number : 1314140-36-7
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-6752
Availability details for
Quantity
  • Availability
  • Add to Cart