COM448619035 Combi-Blocks, Inc.

2-Morpholinothiazole-5-boronic acid, pinacol ester

  • MDL number MFCD13182300
  • Cas Number : 1402172-49-9
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7042

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7042
Availability details for
Quantity
  • Availability
  • Add to Cart