COM448619037 Combi-Blocks, Inc.

1,3,2-Dioxaborolane, 2-[4-(1,1-difluoroethyl)phenyl]-4,4,5,5-tetramethyl-

  • MDL number MFCD13186728
  • Cas Number : 1000994-94-4
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7061

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7061
Availability details for
Quantity
  • Availability
  • Add to Cart