COM448619045 Combi-Blocks, Inc.

Benzothiophene-4-boronic acid pinacol ester

  • MDL number MFCD13619876
  • Cas Number : 1000160-75-7
  • Molecular Weight : 260.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7234

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7234
Availability details for
Quantity
  • Availability
  • Add to Cart