COM448619046 Combi-Blocks, Inc.

3-(Allyloxy)prop-1-en-2-ylboronic acid pinacol ester

  • MDL number MFCD13619879
  • Cas Number : 212127-71-4
  • Molecular Weight : 224.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7238

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7238
Availability details for
Quantity
  • Availability
  • Add to Cart