COM448619050 Combi-Blocks, Inc.

2-Piperidin-1-yl-thiazole-5-boronic acid pinacol ester

  • MDL number MFCD14155981
  • Cas Number : 1350755-62-2
  • Molecular Weight : 294.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7276
Availability details for
Quantity
  • Availability
  • Add to Cart