COM448619057 Combi-Blocks, Inc.

2-Isopentylamino-5-pyridineboronic acid, pinacol ester

  • MDL number MFCD14636461
  • Cas Number : 2096332-90-8
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7360

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7360
Availability details for
Quantity
  • Availability
  • Add to Cart