COM448619063 Combi-Blocks, Inc.

2-Amino-6-chloropyridine-4-boronic acid pinacol ester

  • MDL number MFCD15141957
  • Cas Number : 1558927-43-7
  • Molecular Weight : 254.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7385

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-7385
Availability details for
Quantity
  • Availability
  • Add to Cart