COM448619092 Combi-Blocks, Inc.

2-(4-Fluorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036570
  • Cas Number : 93748-09-5
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-1827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-1827
Availability details for
Quantity
  • Availability
  • Add to Cart