COM448619110 Combi-Blocks, Inc.

Ethyl 4-chloro-2-piperidinopyrimidine-5-carboxylate

  • MDL number MFCD11111793
  • Cas Number : 34750-24-8
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2021
Availability details for
Quantity
  • Availability
  • Add to Cart