COM448619122 Combi-Blocks, Inc.

tert-butyl N-[4-bromo-2-(trifluoromethyl)phenyl]carbamate

  • MDL number MFCD21170461
  • Cas Number : 1393442-28-8
  • Molecular Weight : 340.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2037
Availability details for
Quantity
  • Availability
  • Add to Cart