COM448619131 Combi-Blocks, Inc.

Ethyl 2-(4-(t-BOC)piperazin-1-yl)-4-hydroxypyrimidine-5-carboxylate

  • MDL number MFCD11504885
  • Cas Number : 1065074-31-8
  • Molecular Weight : 352.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2054
Availability details for
Quantity
  • Availability
  • Add to Cart