COM448619132 Combi-Blocks, Inc.

Ethyl 2-(4-(tert-butoxycarbonyl)piperazin-1-yl)-4-methylpyrimidine-5-carboxylate

  • MDL number MFCD12026009
  • Cas Number : 1150163-72-6
  • Molecular Weight : 350.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2055
Availability details for
Quantity
  • Availability
  • Add to Cart