COM448619156 Combi-Blocks, Inc.

Methyl 4-tert-butyl-2-methylthiazole-5-carboxylate

  • MDL number MFCD11504890
  • Cas Number : 1072944-46-7
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2088
Availability details for
Quantity
  • Availability
  • Add to Cart