COM448619159 Combi-Blocks, Inc.

4-(2-Nitrophenoxy)piperidine, HCl

  • MDL number MFCD09909466
  • Cas Number : 1072944-49-0
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2094
Availability details for
Quantity
  • Availability
  • Add to Cart