COM448619161 Combi-Blocks, Inc.

4-Hydroxy-1-(2-nitro-5-trifluoromethylphenyl)piperidine

  • MDL number MFCD11504892
  • Cas Number : 1072944-51-4
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2096
Availability details for
Quantity
  • Availability
  • Add to Cart