COM448619179 Combi-Blocks, Inc.

8-Bromo-6-chloro-3-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD11504898
  • Cas Number : 1072944-56-9
  • Molecular Weight : 276.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2124
Availability details for
Quantity
  • Availability
  • Add to Cart