COM448619180 Combi-Blocks, Inc.

3,8-Dibromo-6-chloroimidazo[1,2-a]pyridine

  • MDL number MFCD11504899
  • Cas Number : 1072944-57-0
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2125
Availability details for
Quantity
  • Availability
  • Add to Cart