COM448619181 Combi-Blocks, Inc.

3,8-Dibromo-6-methylimidazo[1,2-a]pyridine

  • MDL number MFCD11504900
  • Cas Number : 1072944-58-1
  • Molecular Weight : 290.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2126
Availability details for
Quantity
  • Availability
  • Add to Cart